missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenol, TRC

Product Code. 30389348
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389348 Supplier Toronto Research Chemicals Supplier No. TRCI71750525G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Iodophenol
CAS 626-02-8
Molecular Formula C6H5IO
Quantity 25 g
Synonym m-Iodophenol, 3-Iodophenol, m-Hydroxyiodobenzene, m-Iodophenol, m-Iodophenol
InChI Formula InChI=1S/C6H5IO/c7-5-2-1-3-6(8)4-5/h1-4,8H
SMILES Oc1cccc(I)c1
IUPAC Name 3-iodophenol
Molecular Weight (g/mol) 220.01
Formula Weight 219.9385
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.