missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenol, 98%, Thermo Scientific Chemicals

Catalog Number 11489073
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
626-02-8
C6H5IO
220.009
MFCD00002261
FXTKWBZFNQHAAO-UHFFFAOYSA-N
m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968
12272
CHEBI:33439
3-iodophenol
C1=CC(=CC(=C1)I)O
This item is not returnable. View return policy
626-02-8
C6H5IO
220.009
MFCD00002261
FXTKWBZFNQHAAO-UHFFFAOYSA-N
m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968
12272
CHEBI:33439
3-iodophenol
C1=CC(=CC(=C1)I)O

3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 38°C to 43°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2039304
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 3-Iodophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'