missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenol, 98%

Product Code. p-7081253
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489073 10 g 10g
11499073 50 g 50g
11409083 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489073 Supplier Thermo Scientific Alfa Aesar Supplier No. A11155.09
Add to basket
This item is not returnable. View return policy

CAS: 626-02-8 | C6H5IO | 220.009 g/mol

3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-02-8
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.009
MDL Number MFCD00002261
InChI Key FXTKWBZFNQHAAO-UHFFFAOYSA-N
Synonym m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968
PubChem CID 12272
ChEBI CHEBI:33439
IUPAC Name 3-iodophenol
SMILES C1=CC(=CC(=C1)I)O

Specifications

Melting Point 38°C to 43°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2039304
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 3-Iodophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.