missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodophenol, 98%, Thermo Scientific Chemicals
82.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.
Solubility
Slightly soluble in water.
Notes
Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.
Solubility
Slightly soluble in water.
Notes
Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 38°C to 43°C |
Flash Point | >110°C (230°F) |
Quantity | 50 g |
Beilstein | 2039304 |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 220.01 |
Percent Purity | 98% |
Chemical Name or Material | 3-Iodophenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'