missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzylamine, 97%, Thermo Scientific™

£59.20 - £568.90

Chemical Identifiers

CAS 696-40-2
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00192227
InChI Key LQLOGZQVKUNBRX-UHFFFAOYSA-N
Synonym 3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine
PubChem CID 97012
IUPAC Name (3-iodophenyl)methanamine
SMILES C1=CC(=CC(=C1)I)CN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10344782
SDS View Documents Promotion Details
Thermo Scientific Acros
367320010
1g Glass bottle
£59.20
1g
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10417592
SDS View Documents Promotion Details
Thermo Scientific Acros
367320050
5g Glass bottle
£203.80
5g
View alternative products
Save Up to 
»
Estimated Shipment: 25-07-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10398793
SDS View Documents Promotion Details
Thermo Scientific Acros
367320200
20g Glass bottle
£568.90
20g
View alternative products
Save Up to 
»
Estimated Shipment: 25-07-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

696-40-2
233.052
LQLOGZQVKUNBRX-UHFFFAOYSA-N
97012
C1=CC(=CC(=C1)I)CN
C7H8IN
MFCD00192227
3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine
(3-iodophenyl)methanamine

Specifications

3-Iodobenzylamine
96.0
1.7480g/mL
132.0°C (8.0 mmHg)
97%
C7H8IN
MFCD00192227
Glass bottle
3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine
LQLOGZQVKUNBRX-UHFFFAOYSA-N
C1=CC(=CC(=C1)I)CN
233.052
233.05
97%
696-40-2
100.0
Yellow
>110°C
Authentic
IC6H4CH2NH2
12, 2354
1.6380 to 1.6400
1g
1.748
(3-iodophenyl)methanamine
97012
Liquid
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Iodobenzylamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Product Certifications

RUO – Research Use Only