missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzylamine, 97%

Product Code. 11344775
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344775 5 g 5g
11334775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344775 Supplier Thermo Scientific Alfa Aesar Supplier No. L17801.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 696-40-2 | C7H8IN | 233.052 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Air sensitive and light sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-40-2
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00192227
InChI Key LQLOGZQVKUNBRX-UHFFFAOYSA-N
Synonym 3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine
PubChem CID 97012
IUPAC Name (3-iodophenyl)methanamine
SMILES C1=CC(=CC(=C1)I)CN

Specifications

Density 1.748
Boiling Point 132°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6375
Quantity 5 g
UN Number UN2735
Beilstein 2689602
Sensitivity Air and light sensitive
Solubility Information Difficult to mix in water.
Formula Weight 233.05
Percent Purity 97%
Chemical Name or Material 3-Iodobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.