missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzylamine, 97%

Product Code. 10344782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
20 g
Packaging:
Glass bottle
Unit Size:
1g
20g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10344782 1 g Glass bottle 1g
10417592 5 g Glass bottle 5g
10398793 20 g Glass bottle 20g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10344782 Supplier Thermo Scientific Acros Supplier No. 367320010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 696-40-2 | C7H8IN | 233.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-40-2
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00192227
InChI Key LQLOGZQVKUNBRX-UHFFFAOYSA-N
Synonym 3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine
PubChem CID 97012
IUPAC Name (3-iodophenyl)methanamine
SMILES C1=CC(=CC(=C1)I)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.7480g/mL
Boiling Point 132.0°C (8.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula IC6H4CH2NH2
Refractive Index 1.6380 to 1.6400
Quantity 1 g
Beilstein 12, 2354
Specific Gravity 1.748
Formula Weight 233.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Iodobenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Iodobenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.