missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzoyl chloride, 96%, Thermo Scientific™

Product Code. 11318426
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318426 5 g 5g
11308426 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318426 Supplier Thermo Scientific Alfa Aesar Supplier No. L17451.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Reacts with water.

Notes
Moisture and light sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-10-0
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD02093934
InChI Key DTELTOREECFDBC-UHFFFAOYSA-N
PubChem CID 74378
IUPAC Name 3-iodobenzoyl chloride
SMILES C1=CC(=CC(=C1)I)C(=O)Cl

Specifications

Melting Point 23°C to 25°C
Boiling Point 159°C to 160°C (23 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 1860418
Synonym 3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
Formula Weight 266.47
Percent Purity 96%
Chemical Name or Material 3-Iodobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.