missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzotrifluoride, 98+%

Product Code. 11463804
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463804 50 g 50g
11453804 10 g 10g
11473804 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463804 Supplier Thermo Scientific Alfa Aesar Supplier No. A11843.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 401-81-0 | C7H4F3I | 272.009 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-81-0
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00001049
InChI Key IGISPMBUGPHLBY-UHFFFAOYSA-N
Synonym 1-iodo-3-trifluoromethyl benzene, 3-iodobenzotrifluoride, m-iodobenzotrifluoride, benzene, 1-iodo-3-trifluoromethyl, 3-iodobenztrifluoride, m-trifluoromethyl iodobenzene, 3-iodo-alpha,alpha,alpha-trifluorotoluene, 3-trifluoromethyliodobenzene, 3-trifluoromethyl-iodobenzene, alpha,alpha,alpha-trifluoro-m-iodotoluene
PubChem CID 67868
IUPAC Name 1-iodo-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)I)C(F)(F)F

Specifications

Melting Point -8°C
Density 1.882
Boiling Point 81°C to 82°C (24 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.5185
Quantity 50 g
Beilstein 2088596
Sensitivity Light sensitive
Formula Weight 272.01
Percent Purity ≥98%
Chemical Name or Material 3-Iodobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.