missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodobenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Iodobenzoic Acid |
| CAS | 618-51-9 |
| Molecular Formula | C7H5IO2 |
| Quantity | 10 g |
| Synonym | 3-Iodobenzenecarboxylic Acid, NSC 176131, NSC 74693, m-Carboxyiodobenzene, m-Iodobenzoic Acid |
| InChI Formula | InChI=1S/C7H5IO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,(H,9,10) |
| SMILES | OC(=O)c1cccc(I)c1 |
| IUPAC Name | 3-iodobenzoic acid |
| Molecular Weight (g/mol) | 248.02 |
| Formula Weight | 247.9334 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?