missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzeneboronic acid pinacol ester, 97%

Product Code. 11387589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387589 5 g 5g
11377589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387589 Supplier Thermo Scientific Alfa Aesar Supplier No. H28115.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 408492-28-4 | C12H16BIO2 | 329.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 408492-28-4
Molecular Formula C12H16BIO2
Molecular Weight (g/mol) 329.97
MDL Number MFCD09953498
InChI Key HSHFNMSHDHAHDN-UHFFFAOYSA-N
Synonym 2-3-iodophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodophenylboronic acid pinacol ester, amtb088, 3-iodophenylboronic acid pinacolester, 1,3,2-dioxaborolane,2-3-iodophenyl-4,4,5,5-tetramethyl, 2-3-iodanylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodo-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene
PubChem CID 11313483
IUPAC Name 2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1

Specifications

Melting Point 70°C to 72°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 329.97
Percent Purity 97%
Chemical Name or Material 3-Iodobenzeneboronic acid pinacol ester

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.