missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoanisole, 98%

Product Code. 11412297
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412297 5 g 5g
11422297 25 g 25g
11432297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412297 Supplier Thermo Scientific Alfa Aesar Supplier No. A14208.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-85-8 | C7H7IO | 234.036 g/mol

3-Iodoanisole is used to produce 3,3'-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodoanisole is used to produce 3,3′-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in chloroform, alcohol, ether. Insoluble in water.

Notes
Light Sensitive. Incompatible with light and oxidizing agents. Store in the dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-85-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001048
InChI Key RSHBAGGASAJQCH-UHFFFAOYSA-N
Synonym 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole
PubChem CID 69839
IUPAC Name 1-iodo-3-methoxybenzene
SMILES COC1=CC(=CC=C1)I

Specifications

Density 1.78
Boiling Point 243°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.613
Quantity 5 g
Beilstein 1858896
Sensitivity Light sensitive
Solubility Information Soluble in chloroform,alcohol,ether. Insoluble in water.
Formula Weight 234.04
Percent Purity 98%
Chemical Name or Material 3-Iodoanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.