missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoanisole, 97%

Product Code. p-3758224 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10338740

Brand: Thermo Scientific Acros 222020050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-85-8 | C7H7IO | 234.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-85-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.04
MDL Number MFCD00001048
InChI Key RSHBAGGASAJQCH-UHFFFAOYSA-N
Synonym 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole
PubChem CID 69839
IUPAC Name 1-iodo-3-methoxybenzene
SMILES COC1=CC(=CC=C1)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.9600g/mL
Boiling Point 244°C to 245°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4OCH3
Refractive Index 1.6130 to 1.615
Quantity 5 g
Beilstein 06, 208
Specific Gravity 1.96
Formula Weight 234.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Iodoanisole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.