missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoanisole, 97%

Product Code. 10473965 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10473965 100 g Glass bottle 100g
10338740 5 g Glass bottle 5g
10246610 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10473965 Supplier Thermo Scientific Acros Supplier No. 222021000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-85-8 | C7H7IO | 234.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-85-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.04
MDL Number MFCD00001048
InChI Key RSHBAGGASAJQCH-UHFFFAOYSA-N
Synonym 3-iodoanisole, m-iodoanisole, benzene, 1-iodo-3-methoxy, anisole, m-iodo, m-methoxyiodobenzene, 3-methoxyphenyl iodide, 3-methoxyiodobenzene, 1-iodo-3-methoxy-benzene, m-iodoanisol, m-iodo-anisole
PubChem CID 69839
IUPAC Name 1-iodo-3-methoxybenzene
SMILES COC1=CC(=CC=C1)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.9600g/mL
Boiling Point 244°C to 245°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4OCH3
Refractive Index 1.6130 to 1.615
Quantity 100 g
Beilstein 06, 208
Specific Gravity 1.96
Formula Weight 234.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Iodoanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.