missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoaniline 98%, Thermo Scientific™

Product Code. 10520961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10520961 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10520961 Supplier Thermo Scientific Acros Supplier No. 122310250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 626-01-7
CAS Min % 97.5
CAS Max % 100.0
Density 1.8200g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H6IN
Linear Formula IC6H4NH2
MDL Number MFCD00007781
Quantity 25g
Beilstein 12, 670
Synonym m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
InChI Key FFCSRWGYGMRBGD-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)I)N
IUPAC Name 3-iodoaniline
Molecular Weight (g/mol) 219.025
PubChem CID 12271
Formula Weight 219.02
Percent Purity ≥97.5%
Chemical Name or Material 3-Iodoaniline, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.