missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-L-tyrosine, TRC

Product Code. 30137788
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137788 5 g 5g
30133636 10 g 10g
30006536 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137788 Supplier Toronto Research Chemicals Supplier No. TRCI7282505G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-78-0
Molecular Formula C9 H10 I N O3
Molecular Weight (g/mol) 307.09
Synonym L-Tyrosine, 3-iodo-, Tyrosine, 3-iodo-, L- (8CI), 3-Iodo-L-tyrosine, (2S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid, 3-Iodo-4-hydroxyphenylalanine, 3-Iodotyrosine, 4-Hydroxy-3-iodophenylalanine, MIT, MIT (amino acid), NSC 210787, 3-Iodo-L-tyrosine
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
SMILES N[C@@H](Cc1ccc(O)c(I)c1)C(=O)O

Specifications

InChI Formula InChI=1S/C9H10INO3/c10-6-3-5(1-2-8(6)12)4-7(11)9(13)14/h1-3,7,12H,4,11H2,(H,13,14)/t7-/m0/s1
Quantity 5 g
Formula Weight 306.9705
Shipping Condition Room Temperature
Chemical Name or Material 3-Iodo-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.