missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-methylbenzoic acid, 98%

Product Code. 11432277
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432277 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11432277 Supplier Thermo Scientific Alfa Aesar Supplier No. A14197.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82998-57-0 | C8H7IO2 | 262.046 g/mol

3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).

Solubility
Soluble in methanol.

Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82998-57-0
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00010392
InChI Key LDDHMKANNXWUAK-UHFFFAOYSA-N
Synonym 3-iodo-4-methyl-benzoic acid, benzoic acid, 3-iodo-4-methyl, zlchem 953, 3-iodo-p-toluicacid, pubchem3947, 4-carboxy-2-iodotoluene, akos bbb/227, ksc449e9t, 3-iodo-4-methyl benzoic acid, paragos 440021
PubChem CID 621640
IUPAC Name 3-iodo-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)I

Specifications

Melting Point 206°C to 210°C
Quantity 100 g
Beilstein 2436216
Sensitivity Light sensitive
Solubility Information Soluble in methanol.
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material 3-Iodo-4-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.