missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-methylaniline, 98%

Product Code. 11420383
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420383 25 g 25g
11410383 5 g 5g
11430383 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420383 Supplier Thermo Scientific Alfa Aesar Supplier No. A11485.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35944-64-0 | C7H8IN | 233.05 g/mol

3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

Solubility
Insoluble in water.

Notes
Light Sensitive .Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35944-64-0
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.05
MDL Number MFCD00047843
InChI Key RRUDMHNAMZFNEK-UHFFFAOYSA-N
Synonym 3-iodo-p-toluidine, benzenamine, 3-iodo-4-methyl, 4-amino-2-iodotoluene, p-toluidine, 3-iodo, 3-iodo-4-methyl-aniline, benzenamine, 3-iodo-4-methyl-9ci, 3-iodo-4-methyl-phenylamine, pubchem3314, 3-iodo-4-methyl aniline, acmc-209ik4
PubChem CID 118889
IUPAC Name 3-iodo-4-methylaniline
SMILES CC1=CC=C(N)C=C1I

Specifications

Melting Point 34°C to 37°C
Quantity 25 g
UN Number UN2811
Beilstein 2353567
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 233.05
Percent Purity 98%
Chemical Name or Material 3-Iodo-4-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.