missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine

Product Code. 11494390
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494390 1 g 1g
11404400 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494390 Supplier Alfa Aesar™ Supplier No. H50026.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 113975-33-0
Molecular Formula C10H13IN2O
Molecular Weight (g/mol) 304.131
MDL Number MFCD04971323
InChI Key GPMKCDBJLNTANL-UHFFFAOYSA-N
Synonym n-3-iodopyridin-4-yl pivalamide, n-3-iodo-pyridin-4-yl-2,2-dimethyl-propionamide, n-3-iodopyridin-4-yl-2,2-dimethylpropanamide, n-3-iodo-4-pyridinyl-2,2-dimethylpropanamide, 3-iodo-4-2,2,2-trimethylacetamido pyridine, n-3-iodo-4-pyriidinyl-2,2-dimethylpropanamide, propanamide, n-3-iodo-4-pyridinyl-2,2-dimethyl, acmc-20akbz, n-3-iodo-4-pyridyl pivalamide, n-3-iodopyridin-4-yl-2,2-dimethylpropionamide
PubChem CID 819120
IUPAC Name N-(3-iodopyridin-4-yl)-2,2-dimethylpropanamide
SMILES CC(C)(C)C(=O)NC1=C(C=NC=C1)I

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 304.13
Chemical Name or Material 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.