missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl phosphate disodium salt

Product Code. 15423299
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423299 250 mg 250mg
15413299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423299 Supplier Thermo Scientific Alfa Aesar Supplier No. J60552.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A histochemical substrate for alkaline phosphatase | CAS: 3318-43-2 | C8H6NNa2O4P | 257.092 g/mol

A histochemical substrate for alkaline phosphatase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3318-43-2
Molecular Formula C8H6NNa2O4P
Molecular Weight (g/mol) 257.092
MDL Number MFCD00040646
InChI Key WCSKWBKPKXJWEG-UHFFFAOYSA-L
Synonym 3-indoxyl phosphate disodium salt, disodium indol-3-yl phosphate, indol-3-yl sodium phosphate, disodium 1h-indol-3-yl phosphate, unii-p1w2d6403e, indoxylphosphate, disodium salt, sodium 1h-indol-3-yl phosphate, 1h-indol-3-ol, dihydrogen phosphate ester , disodium salt, ip, na2, 3-indoxyl phosphate
PubChem CID 76830
IUPAC Name disodium;1H-indol-3-yl phosphate
SMILES C1=CC=C2C(=C1)C(=CN2)OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Color White
Quantity 250 mg
Formula Weight 257.09
Physical Form Powder
Chemical Name or Material 3-Indoxyl phosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.