missing translation for 'onlineSavingsMsg'
Learn More

3-Indolylacetonitrile, 97%

Product Code. 10453775 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10453775 5 g Glass bottle 5g
10061740 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10453775 Supplier Thermo Scientific Acros Supplier No. 156650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 771-51-7 | C10H8N2 | 156.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-51-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00005628
InChI Key DMCPFOBLJMLSNX-UHFFFAOYSA-N
Synonym 3-indoleacetonitrile, indole-3-acetonitrile, 3-indolylacetonitrile, 2-1h-indol-3-yl acetonitrile, 1h-indole-3-acetonitrile, indolylacetonitrile, 3-cyanomethyl indole, indoleacetonitrile, indolylacetonitril, 3-indolacetonitrile
PubChem CID 351795
ChEBI CHEBI:17566
IUPAC Name 2-(1H-indol-3-yl)acetonitrile
SMILES C1=CC=C2C(=C1)C(=CN2)CC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 35°C to 37°C
Color Colorless to Yellow
Boiling Point 156°C to 160°C (0.2 mmHg)
Flash Point 206°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.6085 to 1.6105
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and toluene
Formula Weight 156.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Indolylacetonitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Indolylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.