missing translation for 'onlineSavingsMsg'
Learn More

3-Indolepropionic acid, MedChemExpress

Product Code. 16423449
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16423449 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16423449 Supplier Medchem Express Supplier No. HYW015229

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress 3-Indolepropionic acid is shown to be a powerful antioxidant and has potential in the treatment for Alzheimer’s disease.

3-Indolepropionic acid is shown to be a powerful antioxidant and has potential in the treatment for Alzheimer’s disease.

Specifications

Chemical Name or Material 3-Indolepropionic acid
CAS 830-96-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H11NO2
Quantity 1 g
Synonym Indole-3-propionic acid 3-IPA
Solubility Information DMSO : ≥ 100 mg/mL (528.51 mM) ∣H2O : 1 mg/mL (5.29 mM; ultrasonic and warming and heat to 80°C)
SMILES C1=CC=CC2=C1C(=C[NH]2)CCC(O)=O
Molecular Weight (g/mol) 189.21
Formula Weight 189.21
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.