missing translation for 'onlineSavingsMsg'
Learn More

3-Indolepropionic acid, 99%, Thermo Scientific Chemicals

Catalog Number 10414395
9.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
830-96-6
C11H11NO2
189.21
MFCD00005660
GOLXRNDWAUTYKT-UHFFFAOYSA-N
3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
3744
CHEBI:43580
3-(1H-indol-3-yl)propanoic acid
OC(=O)CCC1=CNC2=CC=CC=C12
This item is not returnable. View return policy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
830-96-6
C11H11NO2
189.21
MFCD00005660
GOLXRNDWAUTYKT-UHFFFAOYSA-N
3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
3744
CHEBI:43580
3-(1H-indol-3-yl)propanoic acid
OC(=O)CCC1=CNC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 130°C to 136°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 22, 69
Solubility Information Solubility in water: slightly soluble
Formula Weight 189.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Indolepropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Indolepropionic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'