missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytyramine Hydrochloride, 99%

Product Code. 10152680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10152680 10 g Glass bottle 10g
10428850 25 g Glass bottle 25g
10454975 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10152680 Supplier Thermo Scientific Acros Supplier No. 122000100
  • ONLINE EXCLUSIVE PRICE
    £44.10
    £39.65 / 10g
    Save £4.45
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 62-31-7 | C8H12ClNO2 | 189.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-31-7
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012898
InChI Key CTENFNNZBMHDDG-UHFFFAOYSA-N
Synonym dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
PubChem CID 65340
IUPAC Name 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 241°C to 243°C
Color White
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CH2CH2NH2·HCl
Quantity 10 g
Beilstein 13, I, 325
Merck Index 15, 3470
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol and hot 95% ethanol,practically insoluble in ether,chloroform,,benzene and toluene
Formula Weight 189.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxytyramine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.