missing translation for 'onlineSavingsMsg'
Learn More

(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamic Acid 1,1-Dimethylethyl Ester, TRC

Product Code. 30388930
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388930 100 mg
30401595 250 mg
30389540 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388930 Supplier Toronto Research Chemicals Supplier No. TRCH971370100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 847795-89-5
Molecular Formula C15H25NO3
Molecular Weight (g/mol) 267.36
Synonym 1,1-Dimethylethyl N-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamate (ACI), Carbamic acid, (3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)-, 1,1-dimethylethyl ester (9CI), Carbamic acid, N-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)-, 1,1-dimethylethyl ester (ACI)
IUPAC Name tert-butyl N-(3-hydroxy-1-adamantyl)carbamate
SMILES CC(C)(C)OC(=O)NC12CC3CC(CC(O)(C3)C1)C2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H25NO3/c1-13(2,3)19-12(17)16-14-5-10-4-11(6-14)8-15(18,7-10)9-14/h10-11,18H,4-9H2,1-3H3,(H,16,17)
Formula Weight 267.1834
Chemical Name or Material (3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamic Acid 1,1-Dimethylethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.