missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytetrahydrofuran, 97%

Product Code. 10297103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10297103 100 g Glass bottle 100g
10745571 5 g Glass bottle 5g
10578670 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10297103 Supplier Thermo Scientific Acros Supplier No. 121941000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 453-20-3 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 453-20-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00005374
InChI Key XDPCNPCKDGQBAN-UHFFFAOYNA-N
Synonym 3-hydroxytetrahydrofuran, tetrahydrofuran-3-ol, 3-furanol, tetrahydro, tetrahydro-3-furanol, 3-hydroxyoxolane, tetrahydro-furan-3-ol, 3-hydroxytetrahydrofurane, 3-hydroxy tetrahydrofuran, s-3-hydroxytetrahydrofuran, 3-tetrahydrofuranol
PubChem CID 9960
IUPAC Name oxolan-3-ol
SMILES OC1CCOC1

Specifications

CAS Min % 96.0
Color Colorless to Yellow
CAS Max % 100.0
Density 1.0900g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4490 to 1.451
Assay Percent Range 96% min. (GC)
Specific Gravity 1.09
Beilstein 17, 107
Boiling Point 181°C
Formula Weight 88.11
Percent Purity 97%
Chemical Name or Material 3-Hydroxytetrahydrofuran
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxytetrahydrofuran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.