missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytetradecanoic Acid, TRC

Product Code. 30395946
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395946 Supplier Toronto Research Chemicals Supplier No. TRCH9567805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxymyristic Acid
CAS 1961-72-4
Molecular Formula C14 H28 O3
Quantity 5 g
Synonym β-Hydroxymyristic Acid, β-Hydroxytetradecanoic Acid, (3RS)-3-Hydroxytetradecanoic Acid, (RS)-3-Hydroxytetradecanoic Acid, (±)-3-Hydroxytetradecanoic Acid, 3-Hydroxymyristic Acid
InChI Formula InChI=1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-13(15)12-14(16)17/h13,15H,2-12H2,1H3,(H,16,17)
SMILES CCCCCCCCCCCC(O)CC(=O)O
IUPAC Name 3-hydroxytetradecanoic acid
Molecular Weight (g/mol) 244.37
Formula Weight 244.2038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.