missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyrrolidine Hydrochloride, TRC

Product Code. 30402906
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402906 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402906 Supplier Toronto Research Chemicals Supplier No. TRCH953673250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxypyrrolidine Hydrochloride
CAS 86070-82-8
Molecular Formula C4H10ClNO
Quantity 250 mg
Synonym 3-Pyrrolidinol, hydrochloride (1:1) (ACI), 3-Pyrrolidinol, hydrochloride (9CI), 3-Hydroxypyrrolidine hydrochloride, 3-Pyrrolidinol hydrochloride, Pyrrolidin-3-ol hydrochloride
InChI Formula InChI=1S/C4H9NO.ClH/c6-4-1-2-5-3-4;/h4-6H,1-3H2;1H
SMILES Cl.OC1CCNC1
IUPAC Name pyrrolidin-3-ol;hydrochloride
Molecular Weight (g/mol) 123.58
Formula Weight 123.0451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.