missing translation for 'onlineSavingsMsg'
Learn More

(±)-3-Hydroxypyrrolidine hydrochloride, 98%

Product Code. p-7056198
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15492409

Brand: Thermo Scientific Alfa Aesar H64975.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86070-82-8 | C4H10ClNO | 123.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86070-82-8
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD03001754
InChI Key QPMSJEFZULFYTB-UHFFFAOYNA-N
Synonym pyrrolidin-3-ol hydrochloride, 3-hydroxypyrrolidine hydrochloride, 3-pyrrolidinol hydrochloride, 3-hydroxypyrrolidine hcl, 3-pyrrolidinol hcl, 3-pyrrolidinol, hydrochloride, 3-hydroxypyrrolidinehydrochloride, s-3-hydroxypyrrolidine hcl, 3-pyrrolidinol hci, hydroxypyrrolidine hcl
PubChem CID 2769408
IUPAC Name pyrrolidin-3-ol;hydrochloride
SMILES Cl.OC1CCNC1

Specifications

Quantity 5 g
Beilstein 7029051
Formula Weight 123.58
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (±)-3-Hydroxypyrrolidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.