missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine, 98%

Product Code. 10637082 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10637082 500 g Plastic bottle 500g
10122920 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10637082 Supplier Thermo Scientific Acros Supplier No. 121855000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-00-2 | C5H5NO | 95.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-00-2
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.1
MDL Number MFCD00006378
InChI Key GRFNBEZIAWKNCO-UHFFFAOYSA-N
Synonym 3-hydroxypyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-oxopyridine, beta-hydroxypyridine, m-hydroxypyridine, 3-pyridyl alcohol, 3-hydoxypyridine, unii-4kbe4p5b6s
PubChem CID 7971
IUPAC Name pyridin-3-ol
SMILES C1=CC(=CN=C1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 123.0°C to 130.0°C
Color Beige to Brown
Boiling Point 180.0°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 21, 46
Fieser 01,486
Solubility Information Solubility in water: 33g/L water (20°C).
Formula Weight 95.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxypyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.