missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine-2-carboxylic acid, 98%

Product Code. 11371927
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11371927

Brand: Thermo Scientific Alfa Aesar L07489.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 874-24-8 | C6H4NO3 | 138.10 g/mol

3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.

Solubility
Soluble in water (0.25g in 10ml).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-24-8
Molecular Formula C6H4NO3
Molecular Weight (g/mol) 138.10
MDL Number MFCD00006294
InChI Key BRARRAHGNDUELT-UHFFFAOYSA-M
Synonym 3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
PubChem CID 13401
ChEBI CHEBI:64342
IUPAC Name 3-hydroxypyridine-2-carboxylic acid
SMILES OC1=CC=CN=C1C([O-])=O

Specifications

Melting Point ∽214°C (decomposition)
Quantity 5 g
Beilstein 118954
Solubility Information Soluble in water (0.25g in 10ml).
Formula Weight 139.11
Percent Purity 98%
Chemical Name or Material 3-Hydroxypyridine-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.