missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyrazine-2-carboxamide, 98%, Thermo Scientific Chemicals

Product Code. 11397639
Click to view available options
Quantity:
250 mg
1 g
5 g

Apart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Apart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55321-99-8
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00233977
InChI Key SZPBAPFUXAADQV-UHFFFAOYSA-N
Synonym 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry
PubChem CID 294642
IUPAC Name 2-oxo-1H-pyrazine-3-carboxamide
SMILES C1=CN=C(C(=O)N1)C(=O)N

Specifications

Melting Point ∽270°C (decomposition)
Quantity 5 g
Formula Weight 139.12
Percent Purity 98%
Chemical Name or Material 3-Hydroxypyrazine-2-carboxamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'