missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypropionitrile, 97%

Product Code. 11393236
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393236 25 g 25g
11303246 250 g 250g
11313246 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393236 Supplier Thermo Scientific Alfa Aesar Supplier No. L12570.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-78-4 | C3H5NO | 71.079 g/mol

3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-78-4
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.079
MDL Number MFCD00002826
InChI Key WSGYTJNNHPZFKR-UHFFFAOYSA-N
Synonym 3-hydroxypropionitrile, hydracrylonitrile, 2-cyanoethanol, ethylene cyanohydrin, propanenitrile, 3-hydroxy, beta-cyanoethanol, glycol cyanohydrin, methanolacetonitrile, hydroxypropanenitrile, 2-hydroxycyanoethane
PubChem CID 8011
IUPAC Name 3-hydroxypropanenitrile
SMILES C(CO)C#N

Specifications

Melting Point -46°C
Density 1.041
Boiling Point 227°C to 228°C
Flash Point 128°C (262°F)
Linear Formula HO(CH2)2CN
Refractive Index 1.425
Quantity 25 g
Beilstein 635773
Merck Index 14,3794
Solubility Information Fully miscible in water.
Formula Weight 71.08
Percent Purity 98%
Chemical Name or Material 3-Hydroxypropionitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.