missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypromazine, TRC

Product Code. 30394232
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394232 100 mg
30408357 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394232 Supplier Toronto Research Chemicals Supplier No. TRCH952440100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 316-85-8
Molecular Formula C17 H20 N2 O S
Molecular Weight (g/mol) 300.42
Synonym 10H-Phenothiazin-3-ol, 10-[3-(dimethylamino)propyl]-, Phenothiazin-3-ol, 10-[3-(dimethylamino)propyl]- (7CI,8CI), 3-Hydroxypromazine
IUPAC Name 10-[3-(dimethylamino)propyl]phenothiazin-3-ol
SMILES CN(C)CCCN1c2ccccc2Sc3cc(O)ccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H20N2OS/c1-18(2)10-5-11-19-14-6-3-4-7-16(14)21-17-12-13(20)8-9-15(17)19/h3-4,6-9,12,20H,5,10-11H2,1-2H3
Formula Weight 300.1296
Chemical Name or Material 3-Hydroxypromazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.