missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypivalic Acid, TRC

Product Code. 30396691
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396691 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396691 Supplier Toronto Research Chemicals Supplier No. TRCH9501055G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxypivalic Acid
CAS 4835-90-9
Molecular Formula C5H10O3
Quantity 5 g
Synonym 2,2-Dimethyl-3-hydroxypropanoic Acid, 2,2-Dimethyl-3-hydroxypropionic Acid, 2,2-Dimethylhydracrylic Acid, 3-Hydroxy-2,2-dimethylpropanoic Acid, 3-Hydroxy-2,2-dimethylpropionic Acid, Hydroxypivalic Acid, NSC 115936, Oxypivalic Acid, alpha,alpha-Dimethyl-beta-hydroxypropionic Acid
InChI Formula InChI=1S/C5H10O3/c1-5(2,3-6)4(7)8/h6H,3H2,1-2H3,(H,7,8)
SMILES CC(C)(CO)C(=O)O
IUPAC Name 3-hydroxy-2,2-dimethylpropanoic acid
Molecular Weight (g/mol) 118.13
Formula Weight 118.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.