missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypicolinic acid, MedChemExpress

Product Code. 16483459
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483459 Supplier Medchem Express Supplier No. HYY0030
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Hydroxypicolinic acid is a picolinic acid derivative, and belongs to the pyridine family.

3-Hydroxypicolinic acid is a picolinic acid derivative, and belongs to the pyridine family.

Specifications

Chemical Name or Material 3-Hydroxypicolinic acid
CAS 874-24-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H5NO3
Quantity 1 g
Synonym Picolinic acid, 3-hydroxy- (6CI,7CI,8CI) 2-Carboxy-3-hydroxypyridine
Solubility Information DMSO : 65 mg/mL (467.26 mM; Need ultrasonic)
SMILES O=C(C1=NC=CC=C1O)O
Molecular Weight (g/mol) 139.11
Formula Weight 139.11
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.