missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenyltrimethylammonium Iodide, TRC

Product Code. 30395740
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395740 1 g
30404099 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395740 Supplier Toronto Research Chemicals Supplier No. TRCH9512551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2498-27-3
Molecular Formula C9H14INO
Molecular Weight (g/mol) 279.12
Synonym 3-Hydroxy-N,N,N-trimethylbenzenaminium Iodide, (m-Hydroxyphenyl)trimethylammonium Iodide, 3-(Trimethylammonio)phenol Iodide, Trimethyl(3-hydroxyphenyl)ammonium Iodide, m-(Dimethylamino)phenol Methiodide, m-Hydroxy-N-trimethylanilinium Iodide, m-Hydroxytrimethylanilinium Iodide
IUPAC Name (3-hydroxyphenyl)-trimethylazanium;iodide
SMILES C[N+](C)(C)C1=CC(=CC=C1)O.[I-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H13NO.HI/c1-10(2,3)8-5-4-6-9(11)7-8;/h4-7H,1-3H3;1H
Formula Weight 279.012
Chemical Name or Material 3-Hydroxyphenyltrimethylammonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.