missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenyltrimethylammonium Iodide, TRC

Product Code. 30404099
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30404099 10 g
30395740 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30404099 Supplier Toronto Research Chemicals Supplier No. TRCH95125510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 2498-27-3
Molecular Formula C9H14INO
Molecular Weight (g/mol) 279.12
Synonym 3-Hydroxy-N,N,N-trimethylbenzenaminium Iodide, (m-Hydroxyphenyl)trimethylammonium Iodide, 3-(Trimethylammonio)phenol Iodide, Trimethyl(3-hydroxyphenyl)ammonium Iodide, m-(Dimethylamino)phenol Methiodide, m-Hydroxy-N-trimethylanilinium Iodide, m-Hydroxytrimethylanilinium Iodide
IUPAC Name (3-hydroxyphenyl)-trimethylazanium;iodide
SMILES C[N+](C)(C)C1=CC(=CC=C1)O.[I-]

Especificaciones

Quantity 10 g
InChI Formula InChI=1S/C9H13NO.HI/c1-10(2,3)8-5-4-6-9(11)7-8;/h4-7H,1-3H3;1H
Formula Weight 279.012
Chemical Name or Material 3-Hydroxyphenyltrimethylammonium Iodide
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.