missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetylene, 95+%

Product Code. 10186663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10186663 5 g Glass bottle 5g
10390284 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10186663 Supplier Thermo Scientific Acros Supplier No. 392410050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10401-11-3 | C8H6O | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10401-11-3
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00078347
InChI Key AODMJIOEGCBUQL-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci
PubChem CID 139144
IUPAC Name 3-ethynylphenol
SMILES OC1=CC=CC(=C1)C#C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color White to Yellow
Density 1.8030g/mL
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H4C∽CH
Refractive Index 1.5840 to 1.5880 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.803
Formula Weight 118.13
Percent Purity 95+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyphenylacetylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Hydroxyphenylacetylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.