missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetic Acid, TRC

Product Code. 30004163
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004163 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30004163 Supplier Toronto Research Chemicals Supplier No. TRCH9490705G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 621-37-4
Molecular Formula C8 H8 O3
Quantity 5 g
Synonym Benzeneacetic acid, 3-hydroxy-, Acetic acid, (m-hydroxyphenyl)- (7CI,8CI), 3-Hydroxybenzeneacetic acid, (3-Hydroxyphenyl)acetic acid, (m-Hydroxyphenyl)acetic acid, 2-(3-Hydroxyphenyl)acetic acid, 2-(3'-Hydroxyphenyl)acetic acid, 3'-Hydroxyphenylacetic acid, NSC 14360
InChI Formula InChI=1S/C8H8O3/c9-7-3-1-2-6(4-7)5-8(10)11/h1-4,9H,5H2,(H,10,11)
SMILES OC(=O)Cc1cccc(O)c1
IUPAC Name 2-(3-hydroxyphenyl)acetic acid
Molecular Weight (g/mol) 152.15
Formula Weight 152.0473
Chemical Name or Material 3-Hydroxyphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.