missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymorphinan-17-yl]-2-phenyl-ethanone, TRC

Product Code. 30397641
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397641 2.5 mg 2.50mg
30391087 0.5 mg 0.5mg
30398583 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397641 Supplier Toronto Research Chemicals Supplier No. TRCH7846902.5MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxymorphinan-17-yl-2-phenyl-ethanone
Analyte or Component Names 3-Hydroxymorphinan-17-yl-2-phenyl-ethanone
Shipping Condition Room Temperature
Molecular Formula C24 H27 N O2
InChI Formula InChI=1S/C24H27NO2/c26-19-10-9-18-15-22-20-8-4-5-11-24(20,21(18)16-19)12-13-25(22)23(27)14-17-6-2-1-3-7-17/h1-3,6-7,9-10,16,20,22,26H,4-5,8,11-15H2/t20-,22+,24+/m0/s1
Solution Type Neat
SMILES Oc1ccc2C[C@@H]3[C@@H]4CCCC[C@]4(CCN3C(=O)Cc5ccccc5)c2c1
IUPAC Name 1-[(1R,9R,10R)-4-hydroxy-17-azatetracyclo[7.5.3.01,10.02,7]heptadeca-2,4,6-trien-17-yl]-2-phenylethanone
Industry Type Forensics
Molecular Weight (g/mol) 361.477
Quantity 2.5 mg
Formula Weight 361.204
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.