missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymethyl-4-methoxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15499116
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15499116

Brand: Thermo Scientific Alfa Aesar H64738.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 908142-03-0
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD14155851
InChI Key DOQUCBSZQOPLGT-UHFFFAOYSA-N
Synonym 3-hydroxymethyl-4-methoxyphenyl boronic acid, 3-hydroxymethyl-4-methoxyphenylboronic acid, ksc005q4b, 3-hydroxymethyl-4-methoxybenzeneboronic acid, 3-hydroxymethyl-4-methoxyphenyl boronicacid, b-3-hydroxymethyl-4-methoxyphenyl-boronic acid, boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
PubChem CID 53407782
IUPAC Name [3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)OC)CO)(O)O

Specifications

Color White
Quantity 250 mg
Beilstein 15501236
Solubility Information Slightly soluble in water.
Formula Weight 181.98
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Hydroxymethyl-4-methoxybenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.