missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxymethyl-4-methoxybenzeneboronic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important intermediate for organic synthesis and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It is an important intermediate for organic synthesis and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | White |
Quantity | 1 g |
Beilstein | 15501236 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 181.98 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 3-Hydroxymethyl-4-methoxybenzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'