missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymethyl-(1-oxy-2,2,5,5-tetramethylpyrroline), TRC

Product Code. 30408229
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408229 10 mg
30403413 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30408229 Supplier Toronto Research Chemicals Supplier No. TRCH94826010MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55738-75-5
Molecular Formula C9 H16 N O2
Molecular Weight (g/mol) 170.23
Synonym 1H-Pyrrol-1-yloxy, 2,5-dihydro-3-(hydroxymethyl)-2,2,5,5-tetramethyl-, 2,5-Dihydro-3-(hydroxymethyl)-2,2,5,5-tetramethyl-1H-pyrrol-1-yloxy, 3-Hydroxymethyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, 3-Hydroxymethyl-2,2,5,5-tetramethylpyrrolin-1-oxyl, 3-Hydroxymethyl-2,2,5,5-tetramethylpyrroline-N-oxyl, 3-Hydroxymethyl-2,2,5,5-tetramethylpyrroline-1-oxyl
IUPAC Name (1-λ^{1}-oxidanyl-2,2,5,5-tetramethylpyrrol-3-yl)methanol
SMILES CC1(C)C=C(CO)C(C)(C)N1[O]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H16NO2/c1-8(2)5-7(6-11)9(3,4)10(8)12/h5,11H,6H2,1-4H3
Formula Weight 170.1181
Chemical Name or Material 3-Hydroxymethyl-(1-oxy-2,2,5,5-tetramethylpyrroline)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.