missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymandelic Acid, MedChemExpress

Product Code. 16449376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449376 Supplier Medchem Express Supplier No. HYW015326
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Hydroxymandelic Acid, a metabolite of Phenylephrine, Phenylephrine is a α-receptor agonist.

3-Hydroxymandelic Acid, a metabolite of Phenylephrine, Phenylephrine is a α-receptor agonist.

Specifications

Chemical Name or Material 3-Hydroxymandelic Acid
CAS 17119-15-2
Color Light Brown
Purity Grade Notes Research
Molecular Formula C8H8O4
Quantity 100 mg
SMILES O=C(O)C(O)C1=CC=CC(O)=C1
Molecular Weight (g/mol) 168.15
Formula Weight 168.15
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.