missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxylauric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxylauric Acid |
| CAS | 1883-13-2 |
| Molecular Formula | C12 H24 O3 |
| Quantity | 500 mg |
| Synonym | 3-Hydroxy Dodecanoic Acid, (RS)-3-Hydroxylauric Acid, (±)-3-Hydroxydodecanoic Acid, 3-Hydroxydodecanoic Acid, DL-β-Hydroxydodecanoic Acid, β-Hydroxydodecanoic Acid, β-Hydroxylauric Acid |
| InChI Formula | InChI=1S/C12H24O3/c1-2-3-4-5-6-7-8-9-11(13)10-12(14)15/h11,13H,2-10H2,1H3,(H,14,15) |
| SMILES | CCCCCCCCCC(O)CC(=O)O |
| IUPAC Name | 3-hydroxydodecanoic acid |
| Molecular Weight (g/mol) | 216.3172 |
| Formula Weight | 216.1725 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?