missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxylauric Acid, TRC

Product Code. 30396470
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396470 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396470 Supplier Toronto Research Chemicals Supplier No. TRCH94375550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxylauric Acid
CAS 1883-13-2
Molecular Formula C12 H24 O3
Quantity 50 mg
Synonym 3-Hydroxy Dodecanoic Acid, (RS)-3-Hydroxylauric Acid, (±)-3-Hydroxydodecanoic Acid, 3-Hydroxydodecanoic Acid, DL-β-Hydroxydodecanoic Acid, β-Hydroxydodecanoic Acid, β-Hydroxylauric Acid
InChI Formula InChI=1S/C12H24O3/c1-2-3-4-5-6-7-8-9-11(13)10-12(14)15/h11,13H,2-10H2,1H3,(H,14,15)
SMILES CCCCCCCCCC(O)CC(=O)O
IUPAC Name 3-hydroxydodecanoic acid
Molecular Weight (g/mol) 216.3172
Formula Weight 216.1725
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.