missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyisobutyryl CoA, TRC

Product Code. 30400572
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400572 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400572 Supplier Toronto Research Chemicals Supplier No. TRCH9434605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxyisobutyryl CoA
CAS 9025-88-1
Molecular Formula C25 H42 N7 O18 P3 S
Quantity 5 mg
Synonym 3-Hydroxyisobutyryl Coenzyme A Hydrolase, 3-Hydroxyisobutyryl CoA Hydrolase, 3-Hydroxyisobutyryl Coenzyme A, 3-Hydroxyisobutyryl Coenzyme A Hydrolase, E.C. 3.1.2.4, EC 3.1.2.4, β-Hydroxyisobutyryl-CoA Hydrolase, β-Hydroxyisobutyryl-coenzyme A Hydrolase
InChI Formula InChI=1S/C25H42N7O18P3S/c1-13(8-33)24(38)54-7-6-27-15(34)4-5-28-22(37)19(36)25(2,3)10-47-53(44,45)50-52(42,43)46-9-14-18(49-51(39,40)41)17(35)23(48-14)32-12-31-16-20(26)29-11-30-21(16)32/h11-14,17-19,23,33,35-36H,4-10H2,1-3H3,(H,27,34)(H,28,37)(H,42,43)(H,44,45)(H2,26,29,30)(H2,39,40,41)/t13?,14-,17-,18-,19+,23-/m1/s1
SMILES CC(CO)C(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP(=O)(O)OP(=O)(O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23
IUPAC Name S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 3-hydroxy-2-methylpropanethioate
Molecular Weight (g/mol) 853.623
Formula Weight 853.152
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.