missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyhomophthalic Acid, TRC

Product Code. 30399153
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399153 100 mg 100mg
30404376 500 mg -
30407466 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399153 Supplier Toronto Research Chemicals Supplier No. TRCH942843100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42421-12-5
Molecular Formula C9H8O5
Molecular Weight (g/mol) 196.16
Synonym 2,6-Cresotic acid, alpha-carboxy- (7CI), 2-Carboxy-3-hydroxybenzeneacetic acid, 2-(Carboxymethyl)-6-hydroxybenzoic acid
IUPAC Name 2-(carboxymethyl)-6-hydroxybenzoic acid
SMILES OC(=O)Cc1cccc(O)c1C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H8O5/c10-6-3-1-2-5(4-7(11)12)8(6)9(13)14/h1-3,10H,4H2,(H,11,12)(H,13,14)
Formula Weight 196.0372
Chemical Name or Material 3-Hydroxyhomophthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.