missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyhippuric acid, MedChemExpress

Product Code. 16401037
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401037 Supplier Medchem Express Supplier No. HY113085
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 3-Hydroxyhippuric acid is an acyl glycine. Acyl glycines are normally minor metabolites of fatty acids.

3-Hydroxyhippuric acid is an acyl glycine. Acyl glycines are normally minor metabolites of fatty acids.

Specifications

Chemical Name or Material 3-Hydroxyhippuric acid
CAS 1637-75-8
Color White
Purity Grade Notes Research
Molecular Formula C9H9NO4
Solubility Information DMSO : 83.33 mg/mL (426.96 mM; Need ultrasonic)
SMILES O=C(O)CNC(C1=CC=CC(O)=C1)=O
Molecular Weight (g/mol) 195.17
Quantity 50 mg
Formula Weight 195.17
Percent Purity 99.64%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.